2'-Chlor-5'-methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4927-1G
COBBCA-4927-1GEA 864 EUR
COBBCA-4927-1G
2'-Chlor-5'-methoxybiphenyl-3-carbonsäure
2'-Chlor-5'-methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₁ClO₃
MDL: MFCD15143521
CAS-Nummer: 1215205-42-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR