2'-Brom-3'-methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4928-1G
COBBCA-4928-1GEA 864 EUR
COBBCA-4928-1G
2'-Brom-3'-methoxybiphenyl-3-carbonsäure
2'-Brom-3'-methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₁BrO₃
MDL: MFCD15143522
CAS-Nummer: 1215206-07-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR