3'-Brom-5'-methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4930-1G
COBBCA-4930-1GEA 865 EUR
COBBCA-4930-1G
3'-Brom-5'-methoxybiphenyl-3-carbonsäure
3'-Brom-5'-methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₁BrO₃
MDL: MFCD15143524
CAS-Nummer: 1215205-39-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR