2',3'-Dimethoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4932-1G
COBBCA-4932-1GEA 865 EUR
COBBCA-4932-1G
2',3'-Dimethoxybiphenyl-3-carbonsäure
2',3'-Dimethoxybiphenyl-3-carbonsäure
Formel: C₁₅H₁₄O₄
MDL: MFCD15143526
CAS-Nummer: 1215206-70-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR