2'-Fluor-3'-methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4936-1G
COBBCA-4936-1GEA 303 EUR
COBBCA-4936-1G
2'-Fluor-3'-methoxybiphenyl-3-carbonsäure
2'-Fluor-3'-methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₁FO₃
MDL: MFCD15143528
CAS-Nummer: 1215206-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR