2'-Hydroxy-3'-methoxybiphenyl-3-carbonsäure
Lieferant: COMBI-BLOCKS
CA-4940-1G
COBBCA-4940-1GEA
864
EUR
COBBCA-4940-1G
2'-Hydroxy-3'-methoxybiphenyl-3-carbonsäure
2'-Hydroxy-3'-methoxybiphenyl-3-carbonsäure
Formel:
C₁₄H₁₂O₄ |
MDL:
MFCD15143532 CAS-Nummer: 1215205-80-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...