3'-Hydroxy-5'-methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4942-1G
COBBCA-4942-1GEA 864 EUR
COBBCA-4942-1G
3'-Hydroxy-5'-methoxybiphenyl-3-carbonsäure
3'-Hydroxy-5'-methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₂O₄
MDL: MFCD15143534
CAS-Nummer: 1215205-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR