3'-Hydroxy-4'-methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4944-1G
COBBCA-4944-1GEA 864 EUR
COBBCA-4944-1G
3'-Hydroxy-4'-methoxybiphenyl-3-carbonsäure
3'-Hydroxy-4'-methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₂O₄
MDL: MFCD15143536
CAS-Nummer: 1215206-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR