2'-Hydroxy-4'-methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4945-1G
COBBCA-4945-1GEA 983 EUR
COBBCA-4945-1G
2'-Hydroxy-4'-methoxybiphenyl-3-carbonsäure
2'-Hydroxy-4'-methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₂O₄
MDL: MFCD15143537
CAS-Nummer: 1215206-20-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR