2'-Chlor-4'-methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4947-1G
COBBCA-4947-1GEA 204 EUR
COBBCA-4947-1G
2'-Chlor-4'-methoxybiphenyl-3-carbonsäure
2'-Chlor-4'-methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₁ClO₃
MDL: MFCD15143538
CAS-Nummer: 1215206-25-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR