3'-Brom-4'-methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4948-1G
COBBCA-4948-1GEA 76.1 EUR
COBBCA-4948-1G
3'-Brom-4'-methoxybiphenyl-3-carbonsäure
3'-Brom-4'-methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₁BrO₃
MDL: MFCD15143539
CAS-Nummer: 1215206-32-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR