2'-Brom-4'-methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4949-1G
COBBCA-4949-1GEA 864 EUR
COBBCA-4949-1G
2'-Brom-4'-methoxybiphenyl-3-carbonsäure
2'-Brom-4'-methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₁BrO₃
MDL: MFCD15143540
CAS-Nummer: 1215206-38-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR