2'-Fluor-4'-methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4950-1G
COBBCA-4950-1GEA 864 EUR
COBBCA-4950-1G
2'-Fluor-4'-methoxybiphenyl-3-carbonsäure
2'-Fluor-4'-methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₁FO₃
MDL: MFCD12859534
CAS-Nummer: 1182754-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR