3'-Chlor-2'-methylbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4953-1G
COBBCA-4953-1GEA 864 EUR
COBBCA-4953-1G
3'-Chlor-2'-methylbiphenyl-3-carbonsäure
3'-Chlor-2'-methylbiphenyl-3-carbonsäure
Formel: C₁₄H₁₁ClO₂
MDL: MFCD11895768
CAS-Nummer: 1181321-00-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR