6-Brom-o-anisaldehyd

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-6-methoxybenzaldehyde

CA-4957-1G
COBBCA-4957-1GEA 312 EUR
COBBCA-4957-1G
6-Brom-o-anisaldehyd
6-Brom-o-anisaldehyd
Formel: C₈H₇BrO₂
Molecular Weight: 215.05 g/mol
MDL: MFCD08059232
CAS-Nummer: 126712-07-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR