2-(Cyclopentyloxy)-2-phenylessigsäure Natriumsalz

Lieferant: COMBI-BLOCKS

CA-4962-1G
COBBCA-4962-1GEA 623 EUR
COBBCA-4962-1G
2-(Cyclopentyloxy)-2-phenylessigsäure Natriumsalz
2-(Cyclopentyloxy)-2-phenylessigsäure Natriumsalz
Formel: C₁₃H₁₅NaO₃
MDL: MFCD16618938
CAS-Nummer: 39773-76-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR