Methyl-3-brom-5-fluorbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 3-bromo-5-fluorobenzoate

CA-5104-1G
COBBCA-5104-1GEA 36.9 EUR
COBBCA-5104-1G
Methyl-3-brom-5-fluorbenzoat
Methyl-3-brom-5-fluorbenzoat
Formel: C₈H₆BrFO₂
Molecular Weight: 233,04 g/mol
MDL: MFCD07780735
CAS-Nummer: 334792-52-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR