Methyl-3-brom-5-methoxybenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 3-bromo-5-methoxybenzoate

CA-5161-1G
COBBCA-5161-1GEA 570 EUR
COBBCA-5161-1G
Methyl-3-brom-5-methoxybenzoat
Methyl-3-brom-5-methoxybenzoat
Formel: C₉H₉BrO₃
MDL: MFCD11042779
CAS-Nummer: 56709-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR