3,5-Dichlor-4-fluorbenzaldehyd

Lieferant: COMBI-BLOCKS

CA-5440-1G
COBBCA-5440-1GEA 93.7 EUR
COBBCA-5440-1G
3,5-Dichlor-4-fluorbenzaldehyd
3,5-Dichloro-4-fluorobenzaldehyde
Formel: C₇H₃Cl₂FO
Molecular Weight: 193,0000 g/mol
MDL: MFCD11845864
CAS-Nummer: 117820-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR