1-(Phenylsulfonyl)-6-indolboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (1-(Phenylsulphonyl)-1H-indol-6-yl)boronic acid

FA-1082-1G
COBBFA-1082-1GEA 266 EUR
COBBFA-1082-1G
1-(Phenylsulfonyl)-6-indolboronsäure
1-(Phenylsulphonyl)-6-indoleboronic acid
Formel: C₁₄H₁₂BNO₄S
Molecular Weight: 301,1254 g/mol
MDL: MFCD03840611
CAS-Nummer: 480438-52-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR