2-Brom-4-(trifluormethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Bromo-4-(trifluoromethyl)benzeneboronic acid, (2-Bromo-4-(trifluoromethyl)phenyl)boronic acid
FA-1507-1G
COBBFA-1507-1GEA
204
EUR
COBBFA-1507-1G
2-Brom-4-(trifluormethyl)phenylboronsäure
2-Brom-4-(trifluormethyl)phenylboronsäure
Formel:
C₇H₅BBrF₃O₂ Molecular Weight: 268,82 g/mol |
MDL:
MFCD08532500 CAS-Nummer: 959997-88-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...