4-Chlor-2,6-dimethylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Chloro-2,6-dimethylphenyl)boronic acid, 2,6-Dimethyl-4-chlorophenylboronic acid

FA-1763-1G
COBBFA-1763-1GEA 595 EUR
COBBFA-1763-1G
4-Chlor-2,6-dimethylphenylboronsäure
4-Chlor-2,6-dimethylphenylboronsäure
Formel: C₈H₁₀BClO₂
Molecular Weight: 184,4278 g/mol
MDL: MFCD10566594
CAS-Nummer: 1027045-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR