2-Benzyloxy-3-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

FA-2158-1G
COBBFA-2158-1GEA 149 EUR
COBBFA-2158-1G
2-Benzyloxy-3-fluorphenylboronsäure
2-Benzyloxy-3-fluorphenylboronsäure
Formel: C₁₃H₁₂BFO₃
MDL: MFCD11856034
CAS-Nummer: 1256355-53-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR