2-(Methoxymethoxy)-5-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methoxymethoxy-5-(trifluoromethyl)phenylboronic acid

FA-2159-1G
COBBFA-2159-1GEA 204 EUR
COBBFA-2159-1G
2-(Methoxymethoxy)-5-(trifluormethyl)phenylboronsäure
2-(Methoxymethoxy)-5-(trifluormethyl)phenylboronsäure
Formel: C₉H₁₀BF₃O₄
MDL: MFCD11856035
CAS-Nummer: 1256355-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR