4'-Ethylbiphenyl-4-ylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4'-Ethyl-[1,1'-biphenyl]-4-yl)boronic acid

FA-2274-1G
COBBFA-2274-1GEA 58.4 EUR
COBBFA-2274-1G
4'-Ethylbiphenyl-4-ylboronsäure
4'-Ethylbiphenyl-4-ylboronsäure
Formel: C₁₄H₁₅BO₂
Molecular Weight: 226,0800 g/mol
MDL: MFCD04038068
CAS-Nummer: 153035-62-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR