2-Chlor-4-hydroxyphenylboronsäure

Lieferant: COMBI-BLOCKS

FA-2315-5G
COBBFA-2315-5GEA 1310 EUR
COBBFA-2315-5G
2-Chlor-4-hydroxyphenylboronsäure
2-Chlor-4-hydroxyphenylboronsäure
Formel: C₆H₆BClO₃
Molecular Weight: 172,3740 g/mol
MDL: MFCD02684320
CAS-Nummer: 766549-26-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR