4-(Methoxycarbonylmethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(4-(2-Methoxy-2-oxoethyl)phenyl)boronic acid
FA-2401-1G
COBBFA-2401-1GEA
423
EUR
COBBFA-2401-1G
4-(Methoxycarbonylmethyl)phenylboronsäure
4-(Methoxycarbonylmethyl)phenylboronsäure
Formel:
C₉H₁₁BO₄ Molecular Weight: 193,9922 g/mol |
MDL:
MFCD09997796 CAS-Nummer: 454185-96-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...