Benzo[b]furan-3-boronsäure
Lieferant: COMBI-BLOCKS
FA-2484-1G
FA-2484-250M
COBBFA-2484-1GEA
0
EUR
COBBFA-2484-1G
COBBFA-2484-250M
Benzo[b]furan-3-boronsäure
Benzo[b]furan-3-boronsäure
Formel:
C₈H₇BO₃ |
MDL:
MFCD06801687 CAS-Nummer: 317830-83-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...