Piperidin-4-boronsäurepinakolester Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine hydrochloride

FM-2468-1G
COBBFM-2468-1GEA 312 EUR
COBBFM-2468-1G
Piperidin-4-boronsäurepinakolester Hydrochlorid
Piperidin-4-boronsäurepinakolester Hydrochlorid
Formel: C₁₁H₂₃BClNO₂
Molecular Weight: 247,569 g/mol
MDL: MFCD11506073
CAS-Nummer: 1218790-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR