7-Methoxy-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 7-Methoxy-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

FM-2494-1G
COBBFM-2494-1GEA 639 EUR
COBBFM-2494-1G
7-Methoxy-2-indolboronsäurepinakolester
7-Methoxy-2-indolboronsäurepinakolester
Formel: C₁₅H₂₀BNO₃
Molecular Weight: 273,14 g/mol
MDL: MFCD11858357
CAS-Nummer: 1072812-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR