5-Brom-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

FM-2496-1G FM-2496-250M
COBBFM-2496-1GEA 497 EUR
COBBFM-2496-1G COBBFM-2496-250M
5-Brom-2-indolboronsäurepinakolester
5-Brom-2-indolboronsäurepinakolester
Formel: C₁₄H₁₇BBrNO₂
Molecular Weight: 322.0053 g/mol
MDL: MFCD11858360
CAS-Nummer: 1256358-92-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR