5-Methyl-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 5-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

FM-2497-1G
COBBFM-2497-1GEA 595 EUR
COBBFM-2497-1G
5-Methyl-2-indolboronsäurepinakolester
5-Methyl-2-indolboronsäurepinakolester
Formel: C₁₅H₂₀BNO₂
Molecular Weight: 257,1358 g/mol
MDL: MFCD11858361
CAS-Nummer: 1256358-93-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR