6-Fluor-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

FM-2510-1G
COBBFM-2510-1GEA 864 EUR
COBBFM-2510-1G
6-Fluor-2-indolboronsäurepinakolester
6-Fluor-2-indolboronsäurepinakolester
Formel: C₁₄H₁₇BFNO₂
MDL: MFCD11858391
CAS-Nummer: 1256358-98-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR