1-Boc-3-methyl-1H-pyrazol-4-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate

FM-6371-1G
COBBFM-6371-1GEA 1250 EUR
COBBFM-6371-1G
1-Boc-3-methyl-1H-pyrazol-4-boronsäurepinakolester
1-Boc-3-methyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₅H₂₅BN₂O₄
Molecular Weight: 308,18 g/mol
MDL: MFCD12407274
CAS-Nummer: 1009071-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR