7-Chlor-3-nitroimidazo[1,2-a]pyridin
Lieferant: COMBI-BLOCKS
HC-2070-5G
HC-2070-1G
COBBHC-2070-5GEA
884
EUR
COBBHC-2070-5G
COBBHC-2070-1G
7-Chlor-3-nitroimidazo[1,2-a]pyridin
7-Chlor-3-nitroimidazo[1,2-a]pyridin
Formel:
C₇H₄ClN₃O₂ |
MDL:
MFCD11504888 CAS-Nummer: 1072944-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...