2-Amino-4-(4-bromphenyl)thiazol

Lieferant: COMBI-BLOCKS

HC-2098-5G HC-2098-100G HC-2098-25G
COBBHC-2098-5GEA 49.7 EUR
COBBHC-2098-5G COBBHC-2098-100G COBBHC-2098-25G
2-Amino-4-(4-bromphenyl)thiazol
2-Amino-4-(4-bromophenyl)thiazole
Formel: C₉H₇BrN₂S
Molecular Weight: 255.1343 g/mol
MDL: MFCD00051953
CAS-Nummer: 2103-94-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR