8-Chlorchinolin
Lieferant: COMBI-BLOCKS
HC-2105-5G
HC-2105-25G
HC-2105-1G
COBBHC-2105-5GEA
51.3
EUR
COBBHC-2105-5G
COBBHC-2105-25G
COBBHC-2105-1G
8-Chlorchinolin
8-Chloroquinoline
Formel:
C₉H₆ClN Molecular Weight: 163.61 Siedepunkt: 288 Schmelzpunkt: –20 °C Dichte: 1.2834 |
MDL:
MFCD00047618 CAS-Nummer: 611-33-6 EINECS: 210-265-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...