4-(2-Brom-4-nitrophenoxy)tetrahydropyran

Lieferant: COMBI-BLOCKS

HC-2133-5G HC-2133-1G
COBBHC-2133-5GEA 0 EUR
COBBHC-2133-5G COBBHC-2133-1G
4-(2-Brom-4-nitrophenoxy)tetrahydropyran
4-(2-Brom-4-nitrophenoxy)tetrahydropyran
Formel: C₁₁H₁₂BrNO₄
MDL: MFCD11504903
CAS-Nummer: 1072944-62-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR