4-Brom-6-fluor-2-methylchinolin

Lieferant: COMBI-BLOCKS

HC-2160-1G
COBBHC-2160-1GEA 204 EUR
COBBHC-2160-1G
4-Brom-6-fluor-2-methylchinolin
4-Bromo-6-fluoro-2-methylquinoline
Formel: C₁₀H₇BrFN
MDL: MFCD11505132
CAS-Nummer: 1070879-47-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR