4-Brom-7-chlor-2,8-dimethylchinolin

Lieferant: COMBI-BLOCKS

HC-2161-1G
COBBHC-2161-1GEA 204 EUR
COBBHC-2161-1G
4-Brom-7-chlor-2,8-dimethylchinolin
4-Bromo-7-chloro-2,8-dimethylquinoline
Formel: C₁₁H₉BrClN
MDL: MFCD11504910
CAS-Nummer: 1070879-69-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR