4-Brom-2-(trifluormethyl)chinolin

Lieferant: COMBI-BLOCKS

HC-2165-1G
COBBHC-2165-1GEA 470 EUR
COBBHC-2165-1G
4-Brom-2-(trifluormethyl)chinolin
4-Bromo-2-(trifluoromethyl)quinoline
Formel: C₁₀H₅BrF₃N
Molecular Weight: 276,0526 g/mol
MDL: MFCD00277488
CAS-Nummer: 18706-25-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR