4-Brom-7-chlor-8-methyl-2-(trifluormethyl)chinolin

Lieferant: COMBI-BLOCKS

HC-2173-1G
COBBHC-2173-1GEA 204 EUR
COBBHC-2173-1G
4-Brom-7-chlor-8-methyl-2-(trifluormethyl)chinolin
4-Bromo-7-chloro-8-methyl-2-(trifluoromethyl)quinoline
Formel: C₁₁H₆BrClF₃N
MDL: MFCD11504913
CAS-Nummer: 1072944-67-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR