Ethyl-5-amino-1-(4-chlorphenyl)-4-cyan-1H-pyrazol-3-carboxylat
Lieferant: COMBI-BLOCKS
HC-2211-1G
COBBHC-2211-1GEA
204
EUR
COBBHC-2211-1G
Ethyl-5-amino-1-(4-chlorphenyl)-4-cyan-1H-pyrazol-3-carboxylat
Ethyl-5-amino-1-(4-chlorphenyl)-4-cyan-1H-pyrazol-3-carboxylat
Formel:
C₁₃H₁₁ClN₄O₂ |
MDL:
MFCD09835737 CAS-Nummer: 908584-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...