Ethyl-6-chlor-4-hydroxychinolin-3-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: 6-Chloro-4-hydroxyquinoline-3-carboxylic acid ethyl ester, Ethyl 6-chloro-4-hydroxyquinoline-3-carboxylate

HC-2213-1G
COBBHC-2213-1GEA 227 EUR
COBBHC-2213-1G
Ethyl-6-chlor-4-hydroxychinolin-3-carboxylat
Ethyl-6-chlor-4-hydroxychinolin-3-carboxylat
Formel: C₁₂H₁₀ClNO₃
Molecular Weight: 251,6657 g/mol
MDL: MFCD00173342
CAS-Nummer: 70271-77-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR