5-Amino-8-fluorchinolin

Lieferant: COMBI-BLOCKS

Synonyme: 8-Fluoroquinolin-5-amine

HC-2225-1G HC-2225-250M
COBBHC-2225-1GEA 595 EUR
COBBHC-2225-1G COBBHC-2225-250M
5-Amino-8-fluorchinolin
5-Amino-8-fluoroquinoline
Formel: C₉H₇FN₂
Molecular Weight: 162.1637 g/mol
MDL: MFCD09042730
CAS-Nummer: 175229-87-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR