Ethyl-5-amino-4-cyan-1-phenyl-1H-pyrazol-3-carboxylat

Lieferant: COMBI-BLOCKS

HC-2259-1G
COBBHC-2259-1GEA 303 EUR
COBBHC-2259-1G
Ethyl-5-amino-4-cyan-1-phenyl-1H-pyrazol-3-carboxylat
Ethyl-5-amino-4-cyan-1-phenyl-1H-pyrazol-3-carboxylat
Formel: C₁₃H₁₂N₄O₂
MDL: MFCD00197038
CAS-Nummer: 93764-93-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR