2-Brom-6-fluorbenzothiazol

Lieferant: COMBI-BLOCKS

HC-2281-1G
COBBHC-2281-1GEA 188 EUR
COBBHC-2281-1G
2-Brom-6-fluorbenzothiazol
2-Bromo-6-fluorobenzothiazole
Formel: C₇H₃BrFNS
MDL: MFCD07783786
CAS-Nummer: 152937-04-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR