Methyl-5-brom-4-chlor-8-methylchinolin-2-carboxylat

Lieferant: COMBI-BLOCKS

HC-2286-1G
COBBHC-2286-1GEA 204 EUR
COBBHC-2286-1G
Methyl-5-brom-4-chlor-8-methylchinolin-2-carboxylat
Methyl-5-brom-4-chlor-8-methylchinolin-2-carboxylat
Formel: C₁₂H₉BrClNO₂
MDL: MFCD11855876
CAS-Nummer: 1133115-60-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR