Methyl-4,7-dichlor-8-methylchinolin-2-carboxylat

Lieferant: COMBI-BLOCKS

HC-2289-1G
COBBHC-2289-1GEA 204 EUR
COBBHC-2289-1G
Methyl-4,7-dichlor-8-methylchinolin-2-carboxylat
Methyl-4,7-dichlor-8-methylchinolin-2-carboxylat
Formel: C₁₂H₉Cl₂NO₂
MDL: MFCD11855878
CAS-Nummer: 1133115-64-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR