Ethyl-5-amino-1-(3-chlorphenyl)-4-cyan-1H-pyrazol-3-carboxylat

Lieferant: COMBI-BLOCKS

HC-2313-1G
COBBHC-2313-1GEA 149 EUR
COBBHC-2313-1G
Ethyl-5-amino-1-(3-chlorphenyl)-4-cyan-1H-pyrazol-3-carboxylat
Ethyl-5-amino-1-(3-chlorphenyl)-4-cyan-1H-pyrazol-3-carboxylat
Formel: C₁₃H₁₁ClN₄O₂
MDL: MFCD09835738
CAS-Nummer: 96734-87-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR